Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-Oxopiperidine-3-carboxylate Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. M28525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28525G 5 g
M285225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28525G Supplier TCI America Supplier No. M28525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71486-53-8
Molecular Formula C7H12ClNO3
Molecular Weight (g/mol) 193.627
MDL Number MFCD00012769
InChI Key NMAACQILAGCQPR-UHFFFAOYSA-N
Synonym methyl 4-oxopiperidine-3-carboxylate hydrochloride, methyl 4-oxo-3-piperidinecarboxylate hydrochloride, 3-methoxycarbonyl-4-piperidone hydrochloride, methyl 4-oxo-3-piperidinecarboxylate hcl, 3-piperidinecarboxylic acid, 4-oxo-, methyl ester, hydrochloride, 3-methoxycarbonyl-4-oxopiperidinium chloride, methyl 4-oxopiperidine-3-carboxylate, chloride, 4-piperidine carboxylic acid methyl ketone 3-hydrochloride, pubchem12958, methyl 4-oxopiperidine-3-carboxylate hcl
PubChem CID 2724028
IUPAC Name methyl 4-oxopiperidine-3-carboxylate;hydrochloride
SMILES COC(=O)C1CNCCC1=O.Cl

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 5 g
Formula Weight 193.63
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Oxopiperidine-3-carboxylate Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.