accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7149-42-0

IUPAC Name: 1-(1-methylpiperidin-4-yl)methanamine
Synonyms: 1-(1-methylpiperidin-4-yl)methanamine
Molecular Weight (g/mol): 128.22
Molecular Formula: C7H16N2
InChi Key: AGTPSAZJSOQXHJ-UHFFFAOYSA-N
SMILES: CN1CCC(CN)CC1
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)