Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Methyl-4-piperidinyl)methanamine, 97%

Catalog No. AC451460100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451460100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC451460100 Supplier Thermo Scientific Chemicals Supplier No. 451460100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 7149-42-0 | C7H16N2 | 128.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7149-42-0
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.22
MDL Number MFCD05022430
InChI Key AGTPSAZJSOQXHJ-UHFFFAOYSA-N
Synonym 1-methylpiperidin-4-yl methanamine, 1-methyl-piperidin-4-methylamine, 1-methyl-4-piperidinyl methanamine, 4-piperidinemethanamine, 1-methyl, 1-1-methylpiperidin-4-yl methanamine, 1-methyl-4-aminomethyl piperidine, 4-aminomethyl-1-methylpiperidine, c-1-methyl-piperidin-4-yl-methylamine, 1-methylpiperidine-4-methylamine
PubChem CID 81574
IUPAC Name (1-methylpiperidin-4-yl)methanamine
SMILES CN1CCC(CN)CC1

Specifications

Boiling Point 80.0°C to 81.0°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 10 g
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material (1-Methyl-4-piperidinyl)methanamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im
EINECSNumber 230-477-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.