accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONOOH

CAS RN 714971-28-5

IUPAC Name: tert-butyl 3-(hydroxymethyl)morpholine-4-carboxylate
Synonyms: tert-butyl 3-(hydroxymethyl)morpholine-4-carboxylate
Molecular Weight (g/mol): 217.27
Molecular Formula: C10H19NO4
InChi Key: AIQSXVGBMCJQAG-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCOCC1CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results