Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-4-Boc-(3-hydroxymethyl)morpholine, 97%, Thermo Scientific™

Catalog No. AAH6457403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6457403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6457403 Supplier Thermo Scientific Chemicals Supplier No. H6457403

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
An intermediate of BMS-599626, a selective and orally efficacious inhibitor of human epidermal growth factor receptor 1 and 2 kinases.

Solubility
Slightly soluble in water.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 714971-28-5
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.265
MDL Number MFCD06799478
InChI Key AIQSXVGBMCJQAG-QMMMGPOBSA-N
Synonym s-tert-butyl 3-hydroxymethyl morpholine-4-carboxylate, s-4-boc-3-hydroxymethyl morpholine, s-n-boc-3-hydroxymethyl morpholine, tert-butyl 3s-3-hydroxymethyl morpholine-4-carboxylate, s-n-boc-3-hydroxymethylmorpholine, 3 s-hydroxymethyl-4-bocmorpholine, s-4-boc-3-hydroxymethylmorpholine, s-4-boc-3-hydroxymethyl-morpholine, s-3-hydroxymethyl-morpholine-4-carboxylic acid tert-butyl ester, pubchem5634
PubChem CID 1514266
IUPAC Name tert-butyl (3S)-3-(hydroxymethyl)morpholine-4-carboxylate
SMILES CC(C)(C)OC(=O)N1CCOCC1CO

Specifications

Melting Point 80°C (literature)
Color White
Beilstein 10251196
Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 217.27
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (S)-4-Boc-(3-hydroxymethyl)morpholine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.