accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71501-19-4

IUPAC Name: N,N-dimethyl-4-(2,6,7-trihydroxy-3-oxo-3H-xanthen-1-yl)anilinium hydrogen sulfate
Synonyms: N,N-dimethyl-4-(2,6,7-trihydroxy-3-oxoxanthen-1-yl)anilinium hydrogen sulfate
Molecular Weight (g/mol): 461.44
Molecular Formula: C21H19NO9S
InChi Key: YVJPXJIYDRZLQC-UHFFFAOYSA-N
SMILES: OS([O-])(=O)=O.C[NH+](C)C1=CC=C(C=C1)C1=C(O)C(=O)C=C2OC3=CC(O)=C(O)C=C3C=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results