Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-(4'-Dimethylaminophenyl)-2,6,7-trihydroxyfluorone Sulfate Hydrate 96.0+%, TCI America™
SDP

Catalog No. D11181G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D11181G Supplier TCI America Supplier No. D11181G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71501-19-4
Molecular Formula C21H19NO9S
Molecular Weight (g/mol) 461.44
MDL Number MFCD00022273
InChI Key YVJPXJIYDRZLQC-UHFFFAOYSA-N
Synonym Dimethylaminophenylfluorone (so called)
PubChem CID 91659008
IUPAC Name N,N-dimethyl-4-(2,6,7-trihydroxy-3-oxo-3H-xanthen-1-yl)anilinium hydrogen sulfate
SMILES OS([O-])(=O)=O.C[NH+](C)C1=CC=C(C=C1)C1=C(O)C(=O)C=C2OC3=CC(O)=C(O)C=C3C=C12

Specifications

Quantity 1 g
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(4′-Dimethylaminophenyl)-2,6,7-trihydroxyfluorone Sulfate Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.