accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CSSCH3CH3H3C

CAS RN 71522-78-6

IUPAC Name: 2-{[2-(tert-butylsulfanyl)ethyl]sulfanyl}-2-methylpropane
Synonyms: 2-{[2-(tert-butylsulfanyl)ethyl]sulfanyl}-2-methylpropane
Molecular Weight (g/mol): 206.41
Molecular Formula: C10H22S2
InChi Key: HPSWXOHTKYJSJJ-UHFFFAOYSA-N
SMILES: CC(C)(C)SCCSC(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results