Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(tert-butylthio)ethane Borane 90.0+%, TCI America™
SDP

Catalog No. B18271G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B18271G Supplier TCI America Supplier No. B18271G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71522-78-6
Molecular Formula C10H22S2
Molecular Weight (g/mol) 206.41
MDL Number MFCD07776995
InChI Key HPSWXOHTKYJSJJ-UHFFFAOYSA-N
Synonym Borane - 1,2-Bis(tert-butylthio)ethane Complex, S,S′C-Di-tert-butyl-1,2-ethanedithiol Borane
PubChem CID 23106025
IUPAC Name 2-{[2-(tert-butylsulfanyl)ethyl]sulfanyl}-2-methylpropane
SMILES CC(C)(C)SCCSC(C)(C)C

Specifications

Color White
Quantity 1 g
Formula Weight 234.07
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(tert-butylthio)ethane Borane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.