accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7153-22-2

IUPAC Name: ethyl 4-cyanobenzoate
Synonyms: ethyl 4-cyanobenzoate
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: JLSSWDFCYXSLQX-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)