Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-Cyanobenzoate 98.0+%, TCI America™
SDP

Catalog No. E11675G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11675G 5 g
E116725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11675G Supplier TCI America Supplier No. E11675G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7153-22-2
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00001824
InChI Key JLSSWDFCYXSLQX-UHFFFAOYSA-N
Synonym benzoic acid, 4-cyano-, ethyl ester, 4-cyanobenzoic acid ethyl ester, benzoic acid, 4-cyano-,ethyl ester, 4-cyano-benzoic acid ethyl ester, ethyl p-cyanobenzoate, ethyl-4-cyanobenzoate, p-cyano ethyl benzoate, acmc-209ypo, ethyl 4-cyanobenzoate, 4-cyanobenzoic acid, ethyl ester
PubChem CID 81589
IUPAC Name ethyl 4-cyanobenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)C#N

Specifications

Melting Point 55°C
Color White
Boiling Point 140°C
Quantity 5 g
Formula Weight 175.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Cyanobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.