accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3OHO

CAS RN 71556-74-6

IUPAC Name: 3-ethyl-3-(3-hydroxyphenyl)-1-methylazepan-2-one
Synonyms: 3-ethyl-3-(3-hydroxyphenyl)-1-methylazepan-2-one
Molecular Weight (g/mol): 247.34
Molecular Formula: C15H21NO2
InChi Key: YFSJZMANJNLCOQ-UHFFFAOYNA-N
SMILES: CCC1(CCCCN(C)C1=O)C1=CC=CC(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results