Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethyl-3-(3-hydroxyphenyl)-1-methylazepan-2-one ,95%, Thermo Scientific™

Catalog No. BTB13417DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB13417DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB13417DA Supplier Thermo Scientific Chemicals Supplier No. BTB13417DA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71556-74-6
Molecular Formula C15H21NO2
Molecular Weight (g/mol) 247.338
MDL Number MFCD00276840
InChI Key YFSJZMANJNLCOQ-UHFFFAOYSA-N
Synonym 3-ethyl-3-3-hydroxyphenyl-1-methylazepan-2-one, 3-ethylhexahydro-3-3-hydroxyphenyl-1-methyl-2h-azepin-2-one, 3-ethyl-hexahydro-3-3-hydroxyphenyl-1-methyl-2h-azepin-2-one, 3-ethyl-3-hydroxyphenyl-1-methylazepan-2-one, 2h-azepin-2-one,3-ethylhexahydro-3-3-hydroxyphenyl-1-methyl, 3-ethyl-3-3-hydroxy-phenyl-1-methyl-azepan-2-one, hexahydro-3-ethyl-3-3-hydroxyphenyl-1-methyl-2h-azepin-2-one
PubChem CID 2794573
IUPAC Name 3-ethyl-3-(3-hydroxyphenyl)-1-methylazepan-2-one
SMILES CCC1(CCCCN(C1=O)C)C2=CC(=CC=C2)O

Specifications

CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 95%
Quantity 1 g
Formula Weight 247.34
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-Ethyl-3-(3-hydroxyphenyl)-1-methylazepan-2-one
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.