accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(S)(R)H2N

CAS RN 71581-93-6

IUPAC Name: (1R,2S)-N-benzyl-2-(hydroxymethyl)cyclohexan-1-aminium
Synonyms: (1R,2S)-N-benzyl-2-(hydroxymethyl)cyclohexan-1-aminium
Molecular Weight (g/mol): 220.34
Molecular Formula: C14H22NO
InChi Key: BRQFIORUNWWNBM-ZIAGYGMSSA-O
SMILES: OC[C@H]1CCCC[C@H]1[NH2+]CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results