accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHO

CAS RN 716-76-7

IUPAC Name: [1,1'-biphenyl]-3-carboxylic acid
Synonyms: biphenyl-3-carboxylic acid
Molecular Weight (g/mol): 198.22
Molecular Formula: C13H10O2
InChi Key: XNLWJFYYOIRPIO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)