Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biphenyl-3-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. B38505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38505G 5 g
B38501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38505G Supplier TCI America Supplier No. B38505G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 716-76-7
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.221
MDL Number MFCD00045846
InChI Key XNLWJFYYOIRPIO-UHFFFAOYSA-N
Synonym biphenyl-3-carboxylic acid, 3-biphenylcarboxylic acid, 1,1'-biphenyl-3-carboxylic acid, diphenyl-3-carboxylic acid, wln: qvr cr, 3-biphenylcarboxylicacid, m-phenylbenzoic acid, 3-phenyl benzoic acid
PubChem CID 12854
IUPAC Name 3-phenylbenzoic acid
SMILES C1=CC=C(C=C1)C2=CC(=CC=C2)C(=O)O

Specifications

Melting Point 166°C
Color White
Quantity 5 g
Formula Weight 198.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Biphenyl-3-carboxylic Acid
RTECSNumber DV1925000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.