accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOHOOHNH2

CAS RN 71653-06-0

IUPAC Name: 2-amino-3-hydroxybutanedioic acid
Synonyms: 3-hydroxyaspartic acid
Molecular Weight (g/mol): 149.1
Molecular Formula: C4H7NO5
InChi Key: YYLQUHNPNCGKJQ-UHFFFAOYNA-N
SMILES: NC(C(O)C(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results