Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxyaspartic Acid 98.0+%, TCI America™
SDP

Catalog No. H09475G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09475G 5 g
H09471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09475G Supplier TCI America Supplier No. H09475G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71653-06-0
Molecular Formula C4H7NO5
Molecular Weight (g/mol) 149.10
MDL Number MFCD00128126
InChI Key YYLQUHNPNCGKJQ-UHFFFAOYNA-N
Synonym 2-Amino-3-hydroxysuccinic Acid
PubChem CID 5425
ChEBI CHEBI:83981
IUPAC Name 2-amino-3-hydroxybutanedioic acid
SMILES NC(C(O)C(O)=O)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 149.10
Percent Purity ≥98.0% (T,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxyaspartic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.