accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO(R)CH3(S)H3C(S)CH3(S)CH2OOH

CAS RN 71678-03-0

IUPAC Name: 3-{[(1R,2S,4aS,8aS)-1,2,4a-trimethyl-5-methylidene-decahydronaphthalen-1-yl]methyl}-2-hydroxy-5-methoxycyclohexa-2,5-diene-1,4-dione
Synonyms: 3-{[(1R,2S,4aS,8aS)-1,2,4a-trimethyl-5-methylidene-hexahydro-2H-naphthalen-1-yl]methyl}-2-hydroxy-5-methoxycyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol): 358.48
Molecular Formula: C22H30O4
InChi Key: JJWITJNSXCXULM-YVUMSICPSA-N
SMILES: COC1=CC(=O)C(O)=C(C[C@]2(C)[C@@H](C)CC[C@@]3(C)[C@H]2CCCC3=C)C1=O
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results