Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Ilimaquinone

Catalog No. 401250100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-125-0100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-125-0100UG Supplier MilliporeSigma™ Supplier No. 401250100UG
Add to Cart
Add to Cart

A cell-permeable marine sponge metabolite with anti-microbial, anti-HIV, anti-inflammatory, and anti-mitotic properties.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Description 98% by HPLC
Target Proteins Induces a complete and reversible breakdown of Golgi membranes into smaller vesicular structures and causes depolymerization of microtubules.
CAS 71678-03-0
Molecular Formula C22H30O4
Formula Weight 358.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 100 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.