accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7168-54-9

IUPAC Name: 4,4'-diethoxy-1,1'-biphenyl
Synonyms: 4,4'-diethoxy-1,1'-biphenyl
Molecular Weight (g/mol): 242.32
Molecular Formula: C16H18O2
InChi Key: WTLUUYXFCHWUJK-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)C1=CC=C(OCC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results