Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Diethoxybiphenyl 99.0+%, TCI America™
SDP

Catalog No. D11275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D11275G Supplier TCI America Supplier No. D11275G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7168-54-9
Molecular Formula C16H18O2
Molecular Weight (g/mol) 242.32
MDL Number MFCD00026801
InChI Key WTLUUYXFCHWUJK-UHFFFAOYSA-N
Synonym 4,4′C-Biphenetol
PubChem CID 617993
IUPAC Name 4,4'-diethoxy-1,1'-biphenyl
SMILES CCOC1=CC=C(C=C1)C1=CC=C(OCC)C=C1

Specifications

Melting Point 176°C
Color White
Quantity 5 g
Formula Weight 242.32
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diethoxybiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.