accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOO

CAS RN 7170-40-3

IUPAC Name: 5-phenoxypentanoic acid
Synonyms: 5-phenoxypentanoic acid
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: YYEBMOCMHWICRI-UHFFFAOYSA-N
SMILES: OC(=O)CCCCOC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results