Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Phenoxyvaleric Acid 98.0+%, TCI America™
SDP

Catalog No. P13575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13575G Supplier TCI America Supplier No. P13575G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7170-40-3
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD02093487
InChI Key YYEBMOCMHWICRI-UHFFFAOYSA-N
PubChem CID 303586
IUPAC Name 5-phenoxypentanoic acid
SMILES C1=CC=C(C=C1)OCCCCC(=O)O

Specifications

Melting Point 63°C
Color White-Yellow
Boiling Point 165°C
Quantity 5 g
Formula Weight 194.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Phenoxyvaleric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.