accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OClH3C(R)(S)NH3COHN(R)(R)CH3OH(R)O(R)SCH3(R)OH(S)HO(R)HOH

CAS RN 7179-49-9

IUPAC Name: hydrogen (2S,4R)-N-[(1R,2R)-2-hydroxy-1-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(methylsulfanyl)oxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide hydrate chloride
Synonyms: lincomycin
Molecular Weight (g/mol): 461.01
Molecular Formula: C18H37ClN2O7S
InChi Key: LFZGYTBWUHCAKF-DCNJEFSFSA-N
SMILES: [H+].O.[Cl-].CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)O)[C@H]1O[C@H](SC)[C@H](O)[C@@H](O)[C@H]1O
Grade 
  • (1)
  • (2)
Color 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)