accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3H3CH3CCH3SOOO

CAS RN 7182-86-7

IUPAC Name: tetrabutylazanium 4-methylbenzene-1-sulfonate
Synonyms: tetrabutylammonium ion tosylate
Molecular Weight (g/mol): 413.66
Molecular Formula: C23H43NO3S
InChi Key: REAVCZWUMGIGSW-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=C1)S([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results