accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71838-16-9

IUPAC Name: 2-bromo-1-iodo-4-methylbenzene
Synonyms: 2-bromo-1-iodo-4-methylbenzene
Molecular Weight (g/mol): 296.93
Molecular Formula: C7H6BrI
InChi Key: PLAKKSAFIZVHJP-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=C(I)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results