accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3NC

CAS RN 7188-38-7

IUPAC Name: 2-isocyano-2-methylpropane
Synonyms: tert-butyl isocyanide
Molecular Weight (g/mol): 83.13
Molecular Formula: C5H9N
InChi Key: FAGLEPBREOXSAC-UHFFFAOYSA-N
SMILES: CC(C)(C)[N+]#[C-]
Boiling Point 
  • (2)
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)