Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl isocyanide, 97%

Catalog No. AC403600050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC403600050 5 g Glass bottle
AC403600010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC403600050 Supplier Thermo Scientific Chemicals Supplier No. 403600050
  • $209.00
    $167.65 / Each of 1

    Save $41.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7188-38-7 | C5H9N | 83.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7188-38-7
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
MDL Number MFCD00000002
InChI Key FAGLEPBREOXSAC-UHFFFAOYSA-N
Synonym tert-butyl isocyanide, tert-butylisocyanide, t-butylisonitrile, tert-butylisonitrile, tbin, propane, 2-isocyano-2-methyl, 2-methyl-2-isocyanopropane, 2-methylpropyl isocyanide, t-butyl isocyanide, 2-isocyano-2-methyl-propane
PubChem CID 23577
IUPAC Name 2-isocyano-2-methylpropane
SMILES CC(C)(C)[N+]#[C-]

Specifications

Density 0.7300g/mL
Boiling Point 91.0°C
Flash Point −2°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.3740 to 1.3780
Linear Formula (CH3)3CNC
Quantity 5 g
Fieser 02,50; 09,82; 11,99; 12,95; 14,63
Specific Gravity 0.73
Formula Weight 83.13
Percent Purity 97%
Chemical Name or Material tert-Butyl isocyanide, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Fatal if inhaled.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
EINECSNumber 230-553-5
RTECSNumber EQ7102500

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.