accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNO

CAS RN 719-64-2

IUPAC Name: 5-chloro-3-phenyl-2,1-benzoxazole
Synonyms: 5-chloro-3-phenyl-2,1-benzoxazole
Molecular Weight (g/mol): 229.66
Molecular Formula: C13H8ClNO
InChi Key: MUHJZJKVEQASGY-UHFFFAOYSA-N
SMILES: ClC1=CC2=C(ON=C2C=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results