Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-3-phenyl-2,1-benzisoxazole 96.0+%, TCI America™
SDP

Catalog No. C168725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C168725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C168725G Supplier TCI America Supplier No. C168725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 719-64-2
Molecular Formula C13H8ClNO
Molecular Weight (g/mol) 229.663
MDL Number MFCD00014573
InChI Key MUHJZJKVEQASGY-UHFFFAOYSA-N
Synonym 5-chloro-3-phenylbenzo c isoxazole, 5-chloro-3-phenylanthranil, 5-chloro-3-phenyl-2,1-benzisoxazole, 3-phenyl-5-chloroanthranil, pubchem8713, acmc-209okx, maybridge1_004397, 2, 5-chloro-3-phenyl, 5-chloro-3-phenyl-benzo c isoxazole, 3-phenyl-5-chloro-2,1-benzisoxazole
PubChem CID 347291
IUPAC Name 5-chloro-3-phenyl-2,1-benzoxazole
SMILES C1=CC=C(C=C1)C2=C3C=C(C=CC3=NO2)Cl

Specifications

Melting Point 115°C
Quantity 25 g
Formula Weight 229.66
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-3-phenyl-2,1-benzisoxazole
EINECSNumber (5)-3399
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.