accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CS(S)NHOOOHO

CAS RN 71989-28-1

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(methylsulfanyl)butanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid|FMOC-MET-OH
Molecular Weight (g/mol): 371.45
Molecular Formula: C20H21NO4S
InChi Key: BUBGAUHBELNDEW-SFHVURJKSA-N
SMILES: CSCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Grade 
  • (3)
Percent Purity 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (6)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)