Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bromobis(Dimethylamino)Borane

Catalog No. 47666810ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-666-810ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 47-666-810ML Supplier Sigma Aldrich Supplier No. 47666810ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant involved in:Intramolecular cyclizationOxidative addition for synthesis of platinum boryl phosphine bromo complexesSynthesis of fluorenyltripyrazolylborate ligandsPreparation of rigid-backbone conjugated polymersConversion of epoxides to halohydrins

Specifications

CAS 71989-28-1
Melting Point 103°C to 108°C
Molecular Formula C20H23NO3
Linear Formula C20H23NO3
Refractive Index n20/D 1.478
MDL Number MFCD00377789
Quantity 5 g
Synonym 9-Fluorenylmethyl N-(5-hydroxypentyl)carbamate
Molecular Weight (g/mol) 325.4 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.