accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71998-70-4

IUPAC Name: 4-(benzyloxy)butane-1,3-diol
Synonyms: 4-(benzyloxy)butane-1,3-diol
Molecular Weight (g/mol): 196.25
Molecular Formula: C11H16O3
InChi Key: ZVVBUAFZGGHYAO-UHFFFAOYNA-N
SMILES: OCCC(O)COCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results