Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxy-1,3-butanediol 97.0+%, TCI America™
SDP

Catalog No. B33265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B33265G Supplier TCI America Supplier No. B33265G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71998-70-4
Molecular Formula C11H16O3
Molecular Weight (g/mol) 196.246
InChI Key ZVVBUAFZGGHYAO-UHFFFAOYSA-N
PubChem CID 13181679
IUPAC Name 4-phenylmethoxybutane-1,3-diol
SMILES C1=CC=C(C=C1)COCC(CCO)O

Specifications

Melting Point 75°C
Color White-Yellow
Quantity 5 g
Formula Weight 196.25
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxy-1,3-butanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.