accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NONHNH2NHCl

CAS RN 72-40-2

IUPAC Name: 4-amino-1H-imidazole-5-carboxamide hydrochloride
Synonyms: 4-aminoimidazole-5-carboxamide hydrochloride
Molecular Weight (g/mol): 162.58
Molecular Formula: C4H7ClN4O
InChi Key: MXCUYSMIELHIQL-UHFFFAOYSA-N
SMILES: Cl.NC(=O)C1=C(N)N=CN1
Percent Purity 
  • (1)
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)