accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONNClFFF

CAS RN 720-01-4

IUPAC Name: ethyl 4-chloro-2-(trifluoromethyl)pyrimidine-5-carboxylate
Synonyms: ethyl 4-chloro-2-(trifluoromethyl)pyrimidine-5-carboxylate
Molecular Weight (g/mol): 254.59
Molecular Formula: C8H6ClF3N2O2
InChi Key: DSULCDCBENGHAX-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C(N=C1Cl)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results