accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OHFFFFFF

CAS RN 722-92-9

IUPAC Name: 2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
Synonyms: 2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
Molecular Weight (g/mol): 259.15
Molecular Formula: C9H7F6NO
InChi Key: TZEJXCIGVMTMDY-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C(O)(C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)