Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 96%

Catalog No. AAB2404814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2404814 25 g
AAB2404803 1 g
AAB2404806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2404814 Supplier Thermo Scientific Chemicals Supplier No. B2404814
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 722-92-9 | C9H7F6NO | 259.151 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 722-92-9
Molecular Formula C9H7F6NO
Molecular Weight (g/mol) 259.151
MDL Number MFCD00039258
InChI Key TZEJXCIGVMTMDY-UHFFFAOYSA-N
Synonym 2-4-aminophenyl-1,1,1,3,3,3-hexafluoropropan-2-ol, 4-hexafluoro-2-hydroxyisopropyl aniline, 2-4-aminophenyl-1,1,1,3,3,3-hexafluoro-2-propanol, 2-4-aminophenyl hexafluoro-2-propanol, 2-4-amino-phenyl-1,1,1,3,3,3-hexafluoro-propan-2-ol, benzenemethanol,4-amino-a,a-bis trifluoromethyl, enamine_005408, 4-hexafluoro-2-hydroxy-2-propyl aniline, 4-hexafluoro-2-hydroxy-2-propyl-aniline, 2-4-aminophenyl hexafluoropropan-2-ol
PubChem CID 577801
IUPAC Name 2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES C1=CC(=CC=C1C(C(F)(F)F)(C(F)(F)F)O)N

Specifications

Melting Point 147°C to 152°C
Quantity 25 g
Beilstein 1473737
Formula Weight 259.15
Percent Purity 96%
Chemical Name or Material 2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 670-530-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.