accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 72235-52-0

IUPAC Name: (2,4-difluorophenyl)methanaminium
Synonyms: (2,4-difluorophenyl)methanaminium
Molecular Weight (g/mol): 144.14
Molecular Formula: C7H8F2N
InChi Key: QDZZDVQGBKTLHV-UHFFFAOYSA-O
SMILES: [NH3+]CC1=CC=C(F)C=C1F
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)