accessibility menu, dialog, popup

UserName

Viewing profile as user:
PPPdClClFePhPhPhPhPh

CAS RN 72287-26-4

IUPAC Name: Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II)
Synonyms: —
Molecular Weight (g/mol): 731.71
Molecular Formula: C34H28Cl2FeP2Pd
InChi Key: —
SMILES: [Fe].Cl[Pd]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)