Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), Pd 13.5-15.5%, Thermo Scientific Chemicals

Catalog No. AA4650303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4650303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4650303 Supplier Thermo Scientific Chemicals Supplier No. 04650303
Add to Cart

missing translation for 'validMainframeMsg'

Used as a catalyst for Suzuki and Stile couplings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72287-26-4
Molecular Formula C34H28Cl2FeP2Pd
Molecular Weight (g/mol) 731.71
MDL Number MFCD00015757
IUPAC Name Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II)
SMILES [Fe].Cl[Pd]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 266°C to 283°C
Color Orange
Quantity 1 g
Synonym [1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride
Solubility Information Soluble in dichloromethane.
Formula Weight 731.72
Concentration or Composition (by Analyte or Components) Pd 13.5 to 15.5%
Physical Form Solid
Chemical Name or Material Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 460-040-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.