accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CH3CCH3CH3CH3CH3CH3H3CCH3(E)(E)(E)(E)(E)(E)(E)(E)(E)

CAS RN 7235-40-7

IUPAC Name: 1,3,3-trimethyl-2-[(1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-tetramethyl-18-(2,6,6-trimethylcyclohex-1-en-1-yl)octadeca-1,3,5,7,9,11,13,15,17-nonaen-1-yl]cyclohex-1-ene
Synonyms: β-carotene β-karotin all-trans-β-carotene
Molecular Weight (g/mol): 536.89
Molecular Formula: C40H56
InChi Key: OENHQHLEOONYIE-JLTXGRSLSA-N
SMILES: C\C(\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)CCCC1(C)C
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (3)
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (6)
  • (3)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)