Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Carotene Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015360 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
3 x 100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5360 3 x 100 mg
11-101-5359 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5360 Supplier MilliporeSigma Supelco Supplier No. PHR12393X100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 7235-40-7
Physical Form Neat
Linear Formula C40H56
MDL Number MFCD00001556
Synonym beta,beta-Carotene; Provitamin A
Type Certified Reference Material
Quantity 3 x 100 mg
Formula Weight 536.87
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.