accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClNNNNOO

CAS RN 7238-62-2

IUPAC Name: 1-chloro-3-[4-nitro-5-(piperidin-1-yl)-1H-imidazol-1-yl]propan-2-ol
Synonyms: 1-chloro-3-[4-nitro-5-(piperidin-1-yl)imidazol-1-yl]propan-2-ol
Molecular Weight (g/mol): 288.73
Molecular Formula: C11H17ClN4O3
InChi Key: PKWGLVDKUPRXEN-UHFFFAOYNA-N
SMILES: OC(CCl)CN1C=NC(=C1N1CCCCC1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results