Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate, 97%, Thermo Scientific™

Catalog No. MO07617DE
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO07617DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07617DE Supplier Thermo Scientific Chemicals Supplier No. MO07617DE
Add to Cart

Chemical Identifiers

CAS 7238-62-2
Molecular Formula C7H8ClNO2S
Molecular Weight (g/mol) 205.656
InChI Key VUARUZUFHDNJSY-UHFFFAOYSA-N
Synonym ethyl 2-chloro-4-methylthiazole-5-carboxylate, ethyl 2-chloro-4-methyl-5-thiazolecarboxylate, 5-thiazolecarboxylic acid, 2-chloro-4-methyl-, ethyl ester, 2-chloro-4-methyl-5-thiazolecarboxylic acid ethyl ester, 5-thiazolecarboxylicacid, 2-chloro-4-methyl-, ethyl ester, 2-chloro-4-methyl-thiazole-5-carboxylic acid ethyl ester
PubChem CID 594903
IUPAC Name ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)Cl)C

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 205.66
Percent Purity 97%
Chemical Name or Material Ethyl 2-chloro-4-methyl-1, 3-thiazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.