accessibility menu, dialog, popup

UserName

Viewing profile as user:
CuONHNHNN

CAS RN 7239-60-3

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 395.95
Molecular Formula: C20H20CuN4O
InChi Key: LTDJODBAAPGYPQ-UEDAPAAGSA-N
SMILES: [Cu+].O=C1C=CC=C\C1=C\NCCCN\C=C1\C=NC(=N1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results