accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOO

CAS RN 724-98-1

IUPAC Name: 2-(phenoxymethyl)benzoic acid
Synonyms: 2-(phenoxymethyl)benzoic acid
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: YKNORODREYVARM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1COC1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)