Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenoxymethylbenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. P090525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P090525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P090525G Supplier TCI America Supplier No. P090525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 724-98-1
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD00020294
InChI Key YKNORODREYVARM-UHFFFAOYSA-N
Synonym 2-phenoxymethyl benzoic acid, benzoic acid, 2-phenoxymethyl, alpha-phenoxy-o-toluic acid, unii-nw1f23eg53, o-phenoxymethyl benzoic acid, phenoxymethylbenzoic acid, acmc-1bgx0
PubChem CID 69761
IUPAC Name 2-(phenoxymethyl)benzoic acid
SMILES C1=CC=C(C=C1)OCC2=CC=CC=C2C(=O)O

Specifications

Melting Point 126°C
Color White-Yellow
Quantity 25 g
Formula Weight 228.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenoxymethylbenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.