CAS RN 7240-37-1
IUPAC Name:
2,7-diamino-4,6-dimethyl-3-oxo-N1,N9-bis[2,5,9-trimethyl-1,4,7,11,14-pentaoxo-6,13-bis(propan-2-yl)-hexadecahydro-1H-pyrrolo[2,1-i]1-oxa-4,7,10,13-tetraazacyclohexadecan-10-yl]-3H-phenoxazine-1,9-dicarboxamide
Synonyms:
7-aminoactinomycin D
Molecular Weight (g/mol):
1270.45
Molecular Formula:
C62H87N13O16
InChi Key:
YXHLJMWYDTXDHS-UHFFFAOYNA-N
SMILES:
CC(C)C1NC(=O)C(NC(=O)C2=C3N=C4C(OC3=C(C)C(N)=C2)=C(C)C(=O)C(N)=C4C(=O)NC2C(C)OC(=O)C(C(C)C)N(C)C(=O)CN(C)C(=O)C3CCCN3C(=O)C(NC2=O)C(C)C)C(C)OC(=O)C(C(C)C)N(C)C(=O)CN(C)C(=O)C2CCCN2C1=O